Please login to your online account to display your discounted pricing

α,α-Diphenyl-γ-butyrolactone, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  179430050

 View more versions of this product

Catalog No. AC179430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Crystalline Powder
Assay ≥94%
Formula Weight 238.29
Quantity 5g
Color White to brown
EINECS Number 213-499-7
TSCA TSCA
Melting Point 75° to 79°C
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material α, α-Diphenyl-γ-butyrolactone
Infrared Spectrum Authentic
Name Note tech.
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C16H14O2
MDL Number MFCD00005393
CAS 956-89-8
Synonym 4-Hydroxy-2, 2-diphenylbutyric acid γ-lactone

Specifications

Chemical Name or Material α, α-Diphenyl-γ-butyrolactone
Name Note tech.
CAS 956-89-8
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C16H14O2
Formula Weight 238.29
MDL Number MFCD00005393
Synonym 4-Hydroxy-2, 2-diphenylbutyric acid γ-lactone