Please login to your online account to display your discounted pricing

α,α-Diphenyl-γ-butyrolactone, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α, α-Diphenyl-γ-butyrolactone
Name Note tech.
CAS 956-89-8
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C16H14O2
Formula Weight 238.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179430050

  SDS
thermo fisher scientific
179430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Crystalline Powder
Assay ≥94%
Formula Weight 238.29
Quantity 5g
Color White to brown
EINECS Number 213-499-7
TSCA TSCA
Melting Point 75° to 79°C
CAS Max % 100.0
CAS Min % 94.0
Chemical Name or Material α, α-Diphenyl-γ-butyrolactone
Infrared Spectrum Authentic
Name Note tech.
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C16H14O2
MDL Number MFCD00005393
CAS 956-89-8
Synonym 4-Hydroxy-2, 2-diphenylbutyric acid γ-lactone