missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl(4-piperidinyl)methanol, 95%

Manufacturer:  Alfa Aesar™ H6128114

 View more versions of this product

Catalog No. AAH6128114



Specifications

Specifications

Diphenyl(4-piperidinyl)methanol
160°C to 163°C
115-46-8
MFCD00066980
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O
267.372
267.37
95%
White
25g
C18H21NO
13,895
ZMISODWVFHHWNR-UHFFFAOYSA-N
diphenyl(piperidin-4-yl)methanol
15723
Solid

Chemical Identifiers

115-46-8
267.372
ZMISODWVFHHWNR-UHFFFAOYSA-N
15723
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O
C18H21NO
MFCD00066980
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
diphenyl(piperidin-4-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TN0470000

Documents