Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyl(4-piperidinyl)methanol, 95%

Chemical Identifiers

Chemical Name or Material Diphenyl(4-piperidinyl)methanol
CAS 115-46-8
Formula Weight 267.37
MDL Number MFCD00066980
Merck Index 13,895
Synonym α,α-Diphenyl-4-piperidinemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6128114

  SDS
alfa aesar™
H6128114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6128122

  SDS
alfa aesar™
H6128122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.37
Melting Point 160° to 163°C
Chemical Name or Material Diphenyl(4-piperidinyl)methanol
Physical Form Solid
Assay 95%
MDL Number MFCD00066980
CAS 115-46-8
Merck Index 13,895
Synonym α,α-Diphenyl-4-piperidinemethanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TN0470000
Color White
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.