missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenyl-2-pyridylphosphine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427660050

 View more versions of this product

Catalog No. AC427660050


 

Specifications Description & Specifications

Specifications

Crystalline Powder
5g
81° to 86°C
97%
MFCD00192108
74.85 to 80.27% C; 5.17 to 5.55% H; 5.1 to 5.6% N
Authentic
263.28
White to yellow to tan
C17H14NP
37943-90-1
Diphenyl-2-pyridylphosphine
Glass bottle

Chemical Identifiers

97%
263.28
Diphenyl-2-pyridylphosphine
C17H14NP
MFCD00192108

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning