Please login to your online account to display your discounted pricing

Diphenyl-2-pyridylmethane 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC387940050

 View more versions of this product

Catalog No. AC387940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder and Crystals
Formula Weight 245.32
Quantity 5g
Color Light beige to yellow-orange
Melting Point 54° to 61°C
Chemical Name or Material Diphenyl-2-pyridylmethane
Assay Percent Range 98%
Molecular Formula C18H15N
MDL Number MFCD00006309
CAS 3678-70-4
Synonym 2-Diphenylmethylpyridine
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Diphenyl-2-pyridylmethane
CAS 3678-70-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C18H15N
Formula Weight 245.32
MDL Number MFCD00006309
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents