Please login to your online account to display your discounted pricing

Diphenyl-2-pyridylmethane 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diphenyl-2-pyridylmethane
CAS 3678-70-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C18H15N
Formula Weight 245.32
MDL Number MFCD00006309
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387940050

  SDS
fisher scientific acros
AC387940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC387940250

  SDS
thermo fisher scientific
AC387940250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 245.32
Physical Form Crystalline Powder and Crystals
Quantity 5g
Color Light beige to yellow-orange
Melting Point 54° to 61°C
Synonym 2-Diphenylmethylpyridine
Chemical Name or Material Diphenyl-2-pyridylmethane
Molecular Formula C18H15N
MDL Number MFCD00006309
Assay Percent Range 98%
CAS 3678-70-4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.