Please login to your online account to display your discounted pricing

Diphenic anhydride, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: fisher scientific acros  164950500

 View more versions of this product

Catalog No. AC164950500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Flakes
Formula Weight 224.22
Color White to beige or grayish-brown
Quantity 50g
Beilstein 17, 526
EINECS Number 227-950-0
TSCA TSCA
Melting Point 223° to 227°C
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material Diphenic anhydride
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C14H8O3
MDL Number MFCD00004993
CAS 6050-13-1
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Diphenic anhydride
Name Note 98%
CAS 6050-13-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00004993
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents