missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenic anhydride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164950100

 View more versions of this product

Catalog No. AC164950100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diphenic anhydride
Glass bottle
223.0°C to 227.0°C
98%
98%
MFCD00004993
2,2'-biphenyldicarboxylic anhydride
RTSGJTANVBJFFJ-UHFFFAOYSA-N
benzo[d][2]benzoxepine-5,7-dione
72794
Crystalline Powder or Flakes
Authentic
Beige to White or Brown-Gray
10g
6050-13-1
C14H8O3
17, 526
Solubility in water: decomposes.
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O
224.215
224.22
98%

Chemical Identifiers

6050-13-1
224.215
RTSGJTANVBJFFJ-UHFFFAOYSA-N
72794
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O
C14H8O3
MFCD00004993
2,2'-biphenyldicarboxylic anhydride
benzo[d][2]benzoxepine-5,7-dione
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 227-950-0

TSCA : TSCA

SDS Documents