missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenic anhydride, 98%, ACROS Organics™

Chemical Identifiers

CAS 6050-13-1
Assay Percent Range 98%
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00004993
Chemical Name or Material Diphenic anhydride, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC164950100 SDS Thermo Fisher Scientific
164950100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6050-13-1
C14H8O3
MFCD00004993
≥97.5%
98%
224.22
Diphenic anhydride, 98%

Specifications

224.22
Crystalline Powder or Flakes
223° to 227°C
Glass bottle
C14H8O3
6050-13-1
Diphenic anhydride, 98%
White to beige or grayish-brown
10g
17, 526
Authentic
98%
MFCD00004993
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 227-950-0

TSCA : TSCA