Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenic anhydride, 98%

$67.19 - $1,282.26

Chemical Identifiers

Chemical Name or Material Diphenic anhydride
CAS 6050-13-1
Molecular Formula C14H8O3
Formula Weight 224.22
MDL Number MFCD00004993
Beilstein 171255
Synonym 2,2'-Biphenyldicarboxylic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2341509

  SDS
alfa aesar™
B2341509
10g Each for $67.19

AAB2341518

  SDS
alfa aesar™
B2341518
50g Each for $268.20

AAB2341530

  SDS
alfa aesar™
B2341530
250g Each for $1,282.26
Description & Specifications

Specifications

Formula Weight 224.22
Chemical Name or Material Diphenic anhydride
Assay 98%
TSCA TSCA
CAS 6050-13-1
Synonym 2,2'-Biphenyldicarboxylic anhydride
Molecular Formula C14H8O3
Beilstein 171255
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 223° to 228°C
Health Hazard 1 Warning
MDL Number MFCD00004993
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Diphenic anhydride when treated with quinaldine and zinc chloride it produces quinodiphenone. It is used in the determination of fatty alcohol ethoxylates as derivatization reagent. It is used as medicine, dye and plating solution intermediates.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture and oxidizing agents. Protect from humidity and water.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.