Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenic acid, 97%

$84.82 - $354.09

Chemical Identifiers

Chemical Name or Material Diphenic acid
CAS 482-05-3
Molecular Formula C14H10O4
Formula Weight 242.23
MDL Number MFCD00002464
Beilstein 2053625
Merck Index 14,3310
Synonym 2,2'-Biphenyldicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1586318

  SDS
alfa aesar™
A1586318
50g Each for $84.82

AAA1586330

  SDS
alfa aesar™
A1586330
250g Each for $354.09
Description & Specifications

Specifications

Formula Weight 242.23
Merck Index 14,3310
Molecular Formula C14H10O4
TSCA TSCA
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Odorless
Melting Point 226° to 231°C
CAS 482-05-3
Synonym 2,2'-Biphenyldicarboxylic acid
Quantity 50g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00002464
Chemical Name or Material Diphenic acid
Beilstein 2053625

Applications
Diphenic acid is used as a pharmaceutical intermediate and in the synthesis of dyes

Solubility
Sparingly soluble in water (0.1 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.