missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Dipeptidylpeptidase IV, His·Tag™, Human, Recombinant, S. frugiperda
Description
- Recombinant, human DPP IV fused at the N-terminus to a His•Tag™ sequence and expressed in S. frugiperda insect cells
- A serine exopeptidase dimer composed of two identical subunits of 110-130kDa
- DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, and cell-matrix interactions
Specifications
Specifications
| Formulation | In 20mM Tris-HCl, 5mM CaCl₂, 1μM ZnCl₂, 0.05% NaN₃, pH 8.0 |
| Purity | ≥90% by SDS-PAGE |
| Content And Storage | ≤-70°C |
| Form | Liquid |
| Quantity | 5 mIU |
| Synonym | CD26, DPPIV |
| Product Type | Dipeptidylpeptidase IV |