missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dipentene, fraction of terpene hydrocarbons, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390310010

 View more versions of this product

Catalog No. AC390310010


 

Specifications Description & Specifications

Specifications

Liquid
Clear
0.8680g/mL
Dipentene
68956-56-9
1L
Pure
1.4730 to 1.4780 (20°C, 589nm)
MFCD00062992
Glass bottle

Chemical Identifiers

68956-56-9
MFCD00062992
Dipentene
Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger