Please login to your online account to display your discounted pricing

Dipentene, fraction of terpene hydrocarbons, ACROS Organics™

$41.75 - $41.75

Chemical Identifiers

Chemical Name or Material Dipentene
CAS 68956-56-9
Purity Grade Pure
MDL Number MFCD00062992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390310010

  SDS
thermo fisher scientific
AC390310010
Glass bottle 1L Each for $41.75

AC390310025

  SDS
thermo fisher scientific
AC390310025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Quantity 1L
Color Clear
Purity Grade Pure
Density 0.8680g/mL
Chemical Name or Material Dipentene
Refractive Index 1.4730 to 1.4780 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00062992
CAS 68956-56-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.