missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dipentaerythritol, tech. 90%

$28.01 - $87.43

Chemical Identifiers

CAS 126-58-9
Molecular Formula (HOCH2)3CCH2OCH2C(CH2OH)3
Formula Weight 254.28
MDL Number MFCD00004691
Chemical Name or Material Dipentaerythritol
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA187140B SDS Alfa Aesar™
A187140B
1000g Each for $87.43
AAA1871430 SDS Alfa Aesar™
A1871430
250g Each for $28.01
Specifications Description & Specifications

Chemical Identifiers

126-58-9
254.28
Dipentaerythritol
(HOCH2)3CCH2OCH2C(CH2OH)3
MFCD00004691
Technical Grade

Specifications

126-58-9
254.28
215° to 218°C
Dipentaerythritol
Technical Grade
(HOCH2)3CCH2OCH2C(CH2OH)3
MFCD00004691
1000g
90%

Safety and Handling

TSCA : TSCA