Please login to your online account to display your discounted pricing

Dioxane ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Dioxane
CAS 123-91-1
Purity Grade ACS Reagent Grade
Molecular Formula C4H8O2
Formula Weight 88.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK49374

 
avantor performance
493708
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 88.11
Quantity 4L
Packaging Glass
CAS 123-91-1
Chemical Name or Material Dioxane
Molecular Formula C4H8O2