missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-DIOP, 98%

$64.46 - $148.84

Chemical Identifiers

CAS 32305-98-9
Molecular Formula C31H32O2P2
Formula Weight 498.55
MDL Number MFCD00014107
Beilstein 1441045
Synonym (-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
Chemical Name or Material (-)-DIOP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2102903 SDS Alfa Aesar™
B2102903
1g Each for $148.84
AAB21029MD SDS Alfa Aesar™
B21029MD
250mg Each for $64.46
Specifications Description & Specifications

Chemical Identifiers

32305-98-9
498.55
1441045
(-)-DIOP
C31H32O2P2
MFCD00014107
(-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane

Specifications

−12.5° (c=5 in Benzene)
1g
C31H32O2P2
MFCD00014107
(-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane
98%
88° to 90°C
32305-98-9
498.55
1441045
(-)-DIOP

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C