Please login to your online account to display your discounted pricing

Thermo Scientific™ Micro SCX LC Columns

Achieve ion-exclusion separations in columns conveniently pre-assembled with Micro SCX LC Columns for easy connections.

$2,003.05 - $2,090.23

Specifications

Carbon Load -
Column Format Analytical Column, NanoViper
Column Type Ion Exchange
Endcapped -
Max. Pressure 500bar
View More Specs
Products
Catalog Number Mfr. No. Column Format Diameter (Metric) Length (Metric) Particle Size Price Quantity    

DX164565

 
thermo scientific™
164565
Analytical Column, NanoViper 0.3mm 100mm 10µm Each for $2,003.05

DX164566

 
thermo scientific™
164566
Analytical Column, NanoViper 1mm 150mm 5µm Each for $2,090.23
Description & Specifications

Specifications

Carbon Load -
Column Format Analytical Column, NanoViper
Column Type Ion Exchange
Endcapped -
Max. Pressure 500bar
Packing Material Spherical, fully porous ultrapure silica
pH -
Surface Area -
Temperature -
Type Analytical Column, NanoViper
USP Type N/A

Achieve ion-exclusion separations Thermo Scientific™ Micro SCX LC Columns that are conveniently pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings for easy connections. Donnan exclusion causes strong acids to elute in the void volume of the column. Weak acids that are protonated in the acidic eluent are not subject to Donnan exclusion and penetrate into the pores of the resin. Separation is accomplished by differences in pKa, size, and hydrophobicity. In addition, the nanoViper fingertightt UHPLC fitting system provides virtually zero-dead-volume connections.

About the Micro SCX LC Columns UHPLC Fitting System

Supports backpressures up to 100 MPa (1,000 bar/12,600 psi)

Suitable for temperatures up to 80 °C

Makes over-tightening impossible with easy-to-use fingertight design

Eliminates the assembly of Thermo Scientific™ Dionex™ PEEK™ sleeve connections

Works with virtually any valve, and any column, from any manufacturer

Paves the way for the easy, tool-free nano LC setup of any application