Please login to your online account to display your discounted pricing
Thermo Scientific™ DNAPac™ PA100 Oligonucleotide Columns
Features strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides.
$956.54 - $4,205.94
Specifications
Carbon Load -
Column Type Ion Exchange
Endcapped -
pH 2 to 12.5
Stationary Phase Quaternary ammonium
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Particle Size Price Quantity    
DX043010
thermo scientific™
043010
4mm 250mm 13µm Each for $2,068.88
DX043011
thermo scientific™
043011
9mm 250mm 13µm Each for $4,205.94
DXSP2091
thermo scientific™
SP2091
22mm 250mm 13µm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DXSP4016
thermo scientific™
SP4016
2mm 50mm 13µm Each for $1,163.98
DX043018
thermo scientific™
043018
4mm 50mm 13µm Each for $956.54
Description & Specifications
Specifications
Carbon Load -
Column Type Ion Exchange
Endcapped -
pH 2 to 12.5
Stationary Phase Quaternary ammonium
Surface Area -
Temperature 90°C
USP Type N/A
Feature strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides with Thermo Scientific™ Dionex™ DNAPac™ PA100 Oligonucleotide Columns.
  • 13μm pellicular, nonporous polymeric resin with bound quaternary amine-functionalized MicroBeads™
  • Capable of n, n-1 resolution for oligonucleotides
  • Resolves oligonucleotides with secondary structures
  • Compatible with solvent, high pH, and high temperatures
  • Analyzes phosphorothioate-based clinical samples
  • Provides easy scale-up