Please login to your online account to display your discounted pricing

Thermo Scientific Dionex™ Ion Standards


Combined, Flouride, and Sulfate Ion Standards are used for a variety of routine anion, cation, and standard determinations.

Manufacturer: thermo scientific  056933

 View more versions of this product

Catalog No. DX056933

  • / Each
 

Description & Specifications

Specifications

Storage Room Temperature
For Use With Routine anion determinations
Type Standard
Includes Fluoride 20mg/L; Chloride 30mg/L; Nitrite 100mg/L; Bromide 100mg/L; Nitrate 100mg/L; Phosphate 150mg/L; Sulfate 150mg/L
Item Description 50mL
Operation Used for routine anion determinations
Volume (Metric) 50mL
Description Dionex™ Combined Seven Anion Standard I, 50 mL
Standard Components Sodium fluoride, Sodium chloride, Sodium nitrite, Sodium bromide, Sodium nitrate, Sodium phosphate, Sodium sulfate in Water
Standard Concentration 20, 30, 100, 100, 100, 150, 150mg/L

Combined, Flouride, and Sulfate Thermo Scientific™ Dionex™ Ion Standards are used for a variety of routine anion, cation, and standard determinations.