Please login to your online account to display your discounted pricing
Thermo Scientific™ Dionex™ AutoTrace™ 280 Elution Racks
Choose from six elution racks if you need a different size or a replacement. This rack hold tubes or vials for collecting sample extract from the automated SPE system.
$748.97 - $1,125.23
Specifications
Type Elution Rack
View More Specs
Products
Catalog Number Mfr. No. For Use With Price Quantity    
DX071069
thermo scientific™
071069
15mL Conical Tubes Each for $748.97
DX071070
thermo scientific™
071070
16 x 100mm Test Tubes Each for $1,044.85
DX071072
thermo scientific
071072
4mL Screw Cap Vials Each for $1,125.23
Description & Specifications
Specifications
Type Elution Rack
Hold tubes or vials that collect the sample extract from the automated solid phase extraction (SPE) system with Thermo Scientific™ Dionex™ AutoTrace™ 280 Elution Racks.
Six elution racks are available, each holding a different size vial or tube including: 11mm GC vials, 15mL conical tubes, 16x100mm vials, 4mL screw cap vials, and 40mL collection vials. Install the elution rack onto the two pins on the front of the elution station in the Dionex AutoTrace 280 system. Being symmetrical and having no front or back, the eluent rack can be positioned either way. 
About Automated SPE System
The Thermo Scientific™ Dionex™ AutoTrace™ 280 Automated SPE System is ideal for labs doing U.S. EPA methods for water analysis or other methods requiring solid-phase extraction (SPE) sample cleanup. The workstation automatically loads liquid matrices onto an SPE disc or cartridge and elutes the analytes of interest, letting you process up to six samples at a time—with only 15 minutes of operator involvement.
The AutoTrace 280 instrument handles large volume solid-phase extraction of liquid samples from 20mL to 4L. The system can be configured with 1, 3, or 6mL SPE cartridges or 47mm solid-phase disks, using up to five different solvents.