missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals

Wetting and solubilizing agent

Supplier:  Thermo Scientific Chemicals 04420336

 View more versions of this product

Catalog No. AA4420336



Description

Description

Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dioctyl sulfosuccinate sodium salt
153°C to 157°C
C20H37NaO7S
500 g
14,3401
Soluble in water and methanol.
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
444.56
444.55
Solid
577-11-7
1.1 g/mL
MFCD00012455
Hygroscopic
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
23673837
96%

Chemical Identifiers

577-11-7
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
23673837
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
C20H37NaO7S
MFCD00012455
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362

H315-H318

EINECSNumber : 209-406-4

RTECSNumber : WN0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only