missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dioctyl sulfosuccinate sodium salt, 96%

Wetting and solubilizing agent

Manufacturer:  Alfa Aesar™ 4420322

 View more versions of this product

Catalog No. AA4420322



Specifications

Specifications

Dioctyl sulfosuccinate sodium salt
Hygroscopic
100g
C20H37NaO7S
14,3401
Soluble in water and methanol.
CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S(=O)(=O)[O-].[Na+]
444.559
444.55
96%
1.1 g/mL
153°C to 157°C
577-11-7
MFCD00012455
aerosol ot
APSBXTVYXVQYAB-UHFFFAOYSA-M
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
23673837
Solid

Chemical Identifiers

577-11-7
444.559
APSBXTVYXVQYAB-UHFFFAOYSA-M
23673837
CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S(=O)(=O)[O-].[Na+]
C20H37NaO7S
MFCD00012455
aerosol ot
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362

H315-H318

EINECSNumber : 209-406-4

RTECSNumber : WN0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents