Please login to your online account to display your discounted pricing

Dioctyl phthalate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC117091000

 View more versions of this product

Catalog No. AC117091000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 390.55
Quantity 100g
Color Clear
Beilstein 09, IV, 3180
Merck Index 15, 2868
Density 0.9810g/mL
Packaging Glass bottle
Water 0.1% max.
Chemical Name or Material Dioctyl phthalate
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
CAS 117-81-7
Synonym Bis(2-ethylhexyl) phthalate
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Molecular Formula C24H38O4
Additional Information Vapor Pressure: 1.8mbar at 200°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 99%
MDL Number MFCD00009493
Health Hazard 2 GHS H Statement:
May damage fertility. May damage the unborn child.

Specifications

Chemical Name or Material Dioctyl phthalate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C24H38O4
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Formula Weight 390.55
MDL Number MFCD00009493
Synonym Bis(2-ethylhexyl) phthalate
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage fertility. May damage the unborn child.

SDS & Safety Documents