Please login to your online account to display your discounted pricing

Dioctyl phthalate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dioctyl phthalate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C24H38O4
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Formula Weight 390.55
MDL Number MFCD00009493
Synonym Bis(2-ethylhexyl) phthalate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117090250

  SDS
thermo fisher scientific
AC117090250
Metal Drum 25kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC117090050

  SDS
fisher scientific acros
117090050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC117090100

  SDS
thermo fisher scientific
AC117090100
Plastic drum 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 390.55
Quantity 25kg
Color Clear
Beilstein 09, IV, 3180
Merck Index 15, 2868
Packaging Metal Drum
Density 0.9810g/mL
Water 0.1% max.
Synonym Bis(2-ethylhexyl) phthalate
CAS 117-81-7
Additional Information Vapor Pressure: 1.8mbar at 200°C
MDL Number MFCD00009493
Molecular Formula C24H38O4
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Chemical Name or Material Dioctyl phthalate
Assay Percent Range 99%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage fertility. May damage the unborn child.