Please login to your online account to display your discounted pricing

Dioctyl phthalate, 99%, ACROS Organics™


Quantity 2.5kg
Packaging Glass bottle

Manufacturer: fisher scientific acros  117090025

 View more versions of this product

Catalog No. AC117090025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Additional Information Vapor Pressure: 1.8mbar at 200°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Water 0.1% max.
Density 0.9810g/mL
Color Clear
Quantity 2.5kg
Chemical Name or Material Dioctyl phthalate
CAS 117-81-7
Physical Form Clear liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C24H38O4
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Formula Weight 390.55g/mol
MDL Number MFCD00009493
Beilstein 09, IV, 3180
Merck Index 15, 2868
Synonym Bis(2-ethylhexyl) phthalate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage fertility. May damage the unborn child.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dioctyl phthalate
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C24H38O4
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Formula Weight 390.55g/mol
MDL Number MFCD00009493
Synonym Bis(2-ethylhexyl) phthalate
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage fertility. May damage the unborn child.

SDS & Safety Documents