Please login to your online account to display your discounted pricing

Alfa Aesar™ DiO-Lipoprotein, Low Density, Acetylated, Human Plasma

For research use

Specifications

Formulation Liquid
Concentration 200microgram/mL
Recombinant No
Storage Requirements 2° to 8°C
Source Human plasma
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64029AMH

 
alfa aesar™
J64029AMH
200μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formulation Liquid
Concentration 200microgram/mL
Recombinant No
Storage Requirements 2° to 8°C
Source Human plasma
Product Type DiO-Lipoprotein, low density, acetylated, Human Plasma
Format Liquid
Regulatory Status RUO