Please login to your online account to display your discounted pricing

DiMNF, Tocris™

Selective aryl hydrocarbon receptor modulator (SAhRM)

$212.74 - $886.68

Chemical Identifiers

Chemical Name or Material DiMNF
Assay Percent Range >99%
Molecular Formula C21H16O4
Formula Weight 332.35
Synonym 2-(3, 4-Dimethoxyphenyl)-4H-naphtho[1, 2-b]pyran-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

462810

 
r & d systems
RDS462810
10mg Each for $212.74

462850

 
r & d systems
RDS462850
50mg Each for $886.68
Description & Specifications

Specifications

Formula Weight 332.35
Quantity 10mg
Chemical Name or Material DiMNF
Assay Percent Range >99%
Molecular Formula C21H16O4
CAS 14756-24-2
Synonym 2-(3, 4-Dimethoxyphenyl)-4H-naphtho[1, 2-b]pyran-4-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C