Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylzinc, 96%

Chemical Identifiers

Chemical Name or Material Dimethylzinc
CAS 544-97-8
Formula Weight 95.44
MDL Number MFCD00014854
UN Number UN3394
Beilstein 3587195
Merck Index 14,3263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3956314

  SDS
alfa aesar™
3956314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 95.44
Chemical Name or Material Dimethylzinc
Sensitivity Air & Moisture Sensitive
TSCA TSCA
Melting Point −40°C
Physical Form Liquid
Assay 96%
MDL Number MFCD00014854
CAS 544-97-8
UN Number UN3394
Beilstein 3587195
Merck Index 14,3263
Health Hazard 1 Danger
Quantity 25g
Flash Point −18° (−0.4°F)
Health Hazard 2 GHS H Statement
H250-H260-H252-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.386g/mL at 20°C
Boiling Point 46°C
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Packaging Cylinder
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H260-H252-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.