missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyltin oxide

Manufacturer:  Alfa Aesar™ 7116404

 View more versions of this product

Catalog No. AA7116404



Specifications

Specifications

Dimethyltin oxide
2g
MFCD00013838
dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
C[Sn](=O)C
164.779
166.77
2273-45-2
C2H6OSn
UN3146
WNVQCJNZEDLILP-UHFFFAOYSA-N
dimethyl(oxo)tin
75277
Powder

Chemical Identifiers

2273-45-2
164.779
WNVQCJNZEDLILP-UHFFFAOYSA-N
75277
C[Sn](=O)C
C2H6OSn
MFCD00013838
dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
dimethyl(oxo)tin
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

TSCA : TSCA

SDS Documents