missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyltin oxide

Manufacturer:  Alfa Aesar™ 7116409

 View more versions of this product

Catalog No. AA7116409



Specifications

Specifications

Dimethyltin oxide
10g
MFCD00013838
WNVQCJNZEDLILP-UHFFFAOYSA-N
dimethyl(oxo)tin
75277
Powder
2273-45-2
C2H6OSn
UN3146
C[Sn](=O)C
164.779
166.77

Chemical Identifiers

2273-45-2
164.779
WNVQCJNZEDLILP-UHFFFAOYSA-N
dimethyl(oxo)tin
C2H6OSn
MFCD00013838
75277
C[Sn](=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

TSCA : TSCA

SDS Documents