Learn More
Dimethyltin dichloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 347820050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dimethyltin dichloride | |
188.0°C to 190.0°C | |
94% min. (Argentometry) | |
C2H6Cl2Sn | |
MFCD00000517 | |
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity | |
PKKGKUDPKRTKLJ-UHFFFAOYSA-L | |
dichloro(dimethyl)stannane | |
12955 | |
95% |
753-73-1 | |
Authentic | |
Glass bottle | |
(CH3)2Sn(Cl)2 | |
5g | |
Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol | |
C[Sn](C)(Cl)Cl | |
219.66 | |
219.66 |
Chemical Identifiers
753-73-1 | |
219.66 | |
PKKGKUDPKRTKLJ-UHFFFAOYSA-L | |
12955 | |
C[Sn](C)(Cl)Cl |
C2H6Cl2Sn | |
MFCD00000517 | |
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity | |
dichloro(dimethyl)stannane |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 212-039-2
RUO â Research Use Only