Please login to your online account to display your discounted pricing

Dimethyltin dichloride, 95%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  347820010

 View more versions of this product

Catalog No. AC347820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (Argentometry)
Formula Weight 219.66
Physical Form Crystals
Quantity 1g
Color White to pale yellow
Melting Point 103° to 110°C
Linear Formula (CH3)Sn(Cl)
Assay Percent Range 95%
Molecular Formula C2H6Cl2Sn
Chemical Name or Material Dimethyltin dichloride
CAS 753-73-1
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00000517
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dimethyltin dichloride
Assay ≥94% (Argentometry)
Assay Percent Range 95%
Molecular Formula C2H6Cl2Sn
Linear Formula (CH3)Sn(Cl)
Formula Weight 219.66
MDL Number MFCD00000517
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

SDS & Safety Documents