Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyltin dibromide, 98%

Chemical Identifiers

Chemical Name or Material Dimethyltin dibromide
CAS 2767-47-7
Molecular Formula C2H6Br2Sn
Linear Formula (CH3)2SnBr2
Formula Weight 308.57
MDL Number MFCD00013518
UN Number UN3146
Synonym Dibromodimethyltin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1806006

  SDS
alfa aesar™
A1806006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1806014

  SDS
alfa aesar™
A1806014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 308.57
Chemical Name or Material Dimethyltin dibromide
Boiling Point 208° to 213°C
Melting Point 75° to 77°C
Assay 98%
Molecular Formula C2H6Br2Sn
Linear Formula (CH3)2SnBr2
MDL Number MFCD00013518
CAS 2767-47-7
Quantity 5g
UN Number UN3146
Synonym Dibromodimethyltin
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Unpleasant
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
RTECS Number WH6883000

Applications
Dimethyltin dibromide is used as a catalyst in chemical synthesis.

Solubility
It hydrolyzes slowly in water.

Notes
Moisture and air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.