Please login to your online account to display your discounted pricing

Dimethyltin bromide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethyltin bromide
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2 H6 Br2 Sn
Linear Formula (CH3)2SnBr2
Formula Weight 308.59
MDL Number MFCD00013518
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383340250

  SDS
thermo fisher scientific
AC383340250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 308.59
Quantity 25g
Color White
Melting Point 74° to 77°C
Chemical Name or Material Dimethyltin bromide
Assay Percent Range 99%
Molecular Formula C2 H6 Br2 Sn
Linear Formula (CH3)2SnBr2
MDL Number MFCD00013518
CAS 2767-47-7
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.