Please login to your online account to display your discounted pricing

Dimethylsulfate, +99%, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC430831000

 View more versions of this product

Catalog No. AC430831000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 126.13
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Density 1.3300g/mL
Synonym Methyl sulfate
Linear Formula (CH3)2SO4
CAS 77-78-1
Health Hazard 2 GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Chemical Name or Material Dimethyl sulfate
MDL Number MFCD00008416
Molecular Formula C2H6O4S
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Wear eye protection/face protection.
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Dimethyl sulfate
Assay Percent Range 99+%
Molecular Formula C2H6O4S
Linear Formula (CH3)2SO4
Formula Weight 126.13
MDL Number MFCD00008416
Synonym Methyl sulfate
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Wear eye protection/face protection.
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

SDS & Safety Documents