missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylphosphoramidous Dichloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC383350050

 View more versions of this product

Catalog No. AC383350050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Dimethylphosphoramidous dichloride
Colorless to light yellow
683-85-2
96.0% min.
1.5040 to 1.5060 (20°C, 589nm)
97%
(CH3)2NPCl2
MFCD00013613
1.271g/mL
5mL
Authentic
Glass bottle
Liquid
C2H6Cl2NP
145.96

Chemical Identifiers

97%
(CH3)2NPCl2
MFCD00013613
C2H6Cl2NP
145.96
Dimethylphosphoramidous dichloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger