missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylphosphoramidous Dichloride, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C2H6Cl2NP
Linear Formula (CH3)2NPCl2
Formula Weight 145.96
MDL Number MFCD00013613
Chemical Name or Material Dimethylphosphoramidous dichloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC383350050 SDS Thermo Fisher Scientific
AC383350050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC383350250 SDS Thermo Fisher Scientific
AC383350250
Glass bottle 25mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
(CH3)2NPCl2
MFCD00013613
C2H6Cl2NP
145.96
Dimethylphosphoramidous dichloride

Specifications

Liquid
5mL
1.271g/mL
1.5040 to 1.5060 (20°C, 589nm)
683-85-2
96.0% min.
Glass bottle
Dimethylphosphoramidous dichloride
145.96
Colorless to light yellow
(CH3)2NPCl2
C2H6Cl2NP
97%
Authentic
MFCD00013613

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger