missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylphosphinic chloride, 97+%

Manufacturer:  Alfa Aesar™ 3029202

 View more versions of this product

Catalog No. AA3029202



Specifications

Specifications

Dimethylphosphinic chloride
204°C
0.5g
C2H6ClOP
UN3261
CVNMBKFJYRAHPO-UHFFFAOYSA-N
[chloro(methyl)phosphoryl]methane
517954
Crystalline
Moisture Sensitive
68°C
1111-92-8
MFCD00002078
dimethylphosphinic chloride, phosphinic chloride, dimethyl, dimethylphosphinoyl chloride, phosphinic chloride,dimethyl-6ci,7ci,8ci,9ci, me2p o cl, dimethylphosphinoylchloride, dimethylphosphinyl chloride, acmc-1bo3s, dimethylphosphin1c chloride, dimethyl phosphinyl chloride
CP(=O)(C)Cl
112.493
112.51
≥97%

Chemical Identifiers

1111-92-8
112.493
CVNMBKFJYRAHPO-UHFFFAOYSA-N
517954
CP(=O)(C)Cl
C2H6ClOP
MFCD00002078
dimethylphosphinic chloride, phosphinic chloride, dimethyl, dimethylphosphinoyl chloride, phosphinic chloride,dimethyl-6ci,7ci,8ci,9ci, me2p o cl, dimethylphosphinoylchloride, dimethylphosphinyl chloride, acmc-1bo3s, dimethylphosphin1c chloride, dimethyl phosphinyl chloride
[chloro(methyl)phosphoryl]methane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents