Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylphenylsilanol, 97%

$93.42 - $93.42

Chemical Identifiers

Chemical Name or Material Dimethylphenylsilanol
CAS 5272-18-4
Molecular Formula C8H12OSi
Formula Weight 152.27
MDL Number MFCD08276879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2737703

  SDS
alfa aesar™
H2737703
1g Each for $93.42
Description & Specifications

Specifications

Formula Weight 152.27
CAS 5272-18-4
Molecular Formula C8H12OSi
TSCA TSCA
MDL Number MFCD08276879
Assay 97%
Quantity 1g
Chemical Name or Material Dimethylphenylsilanol
Boiling Point 60° to 62°C (0.5mmHg)