Please login to your online account to display your discounted pricing

Dimethylphenylsilane, 98%, ACROS Organics™

$31.72 - $31.72

Chemical Identifiers

Chemical Name or Material Dimethylphenylsilane
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H12Si
Linear Formula C6H5SiH(CH3)2
Formula Weight 136.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215930050

  SDS
thermo fisher scientific
AC215930050
Glass bottle 5g Each for $31.72
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 136.27
Quantity 5g
Color Clear colorless
Density 0.8890g/mL
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Chemical Name or Material Dimethylphenylsilane
Molecular Formula C8H12Si
CAS 766-77-8
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula C6H5SiH(CH3)2
Health Hazard 1 Warning
Infrared Spectrum Authentic

Reduction reagent

Safety and Handling

Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.