Please login to your online account to display your discounted pricing

Dimethylphenylsilane, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC215930050

 View more versions of this product

Catalog No. AC215930050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 136.27
Quantity 5g
Color Clear colorless
Density 0.8890g/mL
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Chemical Name or Material Dimethylphenylsilane
Molecular Formula C8H12Si
CAS 766-77-8
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula C6H5SiH(CH3)2
Health Hazard 1 Warning
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Dimethylphenylsilane
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H12Si
Linear Formula C6H5SiH(CH3)2
Formula Weight 136.27
Accessories

Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.

SDS & Safety Documents