Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyloxaloylglycine, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67077MA

 View more versions of this product

Catalog No. AAJ67077MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.14
Chemical Name or Material Dimethyloxaloylglycine
Physical Form Solid
Assay 98%
MDL Number MFCD05865098
CAS 89464-63-1
Synonym DMOG; N-(Methoxyoxoacetyl)glycine methyl ester
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Health Hazard 1 Warning
Color Off-white
Quantity 10mg

Specifications

Chemical Name or Material Dimethyloxaloylglycine
CAS 89464-63-1
Formula Weight 175.14
MDL Number MFCD05865098
Synonym DMOG; N-(Methoxyoxoacetyl)glycine methyl ester
Accessories

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

-30° to -10°C