missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylmethoxy(3,3,3-trifluoropropyl)silane, 98%

$58.92 - $200.47

Chemical Identifiers

CAS 4852-13-5
Molecular Formula C6H13F3OSi
Molecular Weight (g/mol) 186.249
MDL Number MFCD00236726
InChI Key QRLCLBZGWYJTHP-UHFFFAOYSA-N
Synonym dimethylmethoxy 3,3,3-trifluoropropyl silane, methoxydimethyl 3,3,3-trifluoropropyl silane, acmc-20al5w, 3,3,3-trifluoropropyl dimethyl methoxysilane, 3,3,3-trifluoropropyl dimethyl methoxysilane, methoxy dimethyl 3,3,3-trifluoropropyl silane
PubChem CID 22497830
IUPAC Name methoxy-dimethyl-(3,3,3-trifluoropropyl)silane
SMILES CO[Si](C)(C)CCC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1764203 View Documents Alfa Aesar™
L1764203
1g Each for $58.92
 
AAL1764206 View Documents Alfa Aesar™
L1764206
5g Each for $200.47
 
Specifications

Chemical Identifiers

4852-13-5
186.249
QRLCLBZGWYJTHP-UHFFFAOYSA-N
22497830
CO[Si](C)(C)CCC(F)(F)F
C6H13F3OSi
MFCD00236726
dimethylmethoxy 3,3,3-trifluoropropyl silane, methoxydimethyl 3,3,3-trifluoropropyl silane, acmc-20al5w, 3,3,3-trifluoropropyl dimethyl methoxysilane, 3,3,3-trifluoropropyl dimethyl methoxysilane, methoxy dimethyl 3,3,3-trifluoropropyl silane
methoxy-dimethyl-(3,3,3-trifluoropropyl)silane

Specifications

4852-13-5
MFCD00236726
dimethylmethoxy 3,3,3-trifluoropropyl silane, methoxydimethyl 3,3,3-trifluoropropyl silane, acmc-20al5w, 3,3,3-trifluoropropyl dimethyl methoxysilane, 3,3,3-trifluoropropyl dimethyl methoxysilane, methoxy dimethyl 3,3,3-trifluoropropyl silane
CO[Si](C)(C)CCC(F)(F)F
186.249
186.25
1g
C6H13F3OSi
UN1993
QRLCLBZGWYJTHP-UHFFFAOYSA-N
methoxy-dimethyl-(3,3,3-trifluoropropyl)silane
22497830
98%
Dimethylmethoxy(3,3,3-trifluoropropyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.