missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylglyoxime, Reagent ACS, +99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 408335000

 View more versions of this product

Catalog No. AC408335000


 

Specifications Description & Specifications

Specifications

Dimethylglyoxime
White to off-white
500g
99.0% min.
0.05% max. (in alcohol)
Crystalline Powder
Plastic bottle
CH3C(=NOH)C(=NOH)CH3
MFCD00002117
0.05% max.
(for nickel determination) passes
>70°C
≈ 240°C (dec.)
95-45-4
Authentic
ACS Reagent Grade
99+%
C4H8N2O2
116.12
01, 772
15, 3270
2, 3-Butanedione dioxime

Chemical Identifiers

95-45-4
C4H8N2O2
116.12
2, 3-Butanedione dioxime
ACS Reagent Grade
99+%
CH3C(=NOH)C(=NOH)CH3
MFCD00002117
Dimethylglyoxime

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-420-1

RTECSNumber : EK2975000

TSCA : TSCA