Please login to your online account to display your discounted pricing

Dimethylglyoxime, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  193804

 View more versions of this product

Catalog No. 02002747

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 116.12
Chemical Name or Material Dimethylglyoxime
Physical Form Powder
Molecular Formula CH3C(:NOH)C(:NOH)CH3
CAS 95-45-4
Quantity 125g
Packaging Poly Bottle
Specific Gravity 1L = 1.37kg

Specifications

Chemical Name or Material Dimethylglyoxime
CAS 95-45-4
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3C(:NOH)C(:NOH)CH3
Formula Weight 116.12