Please login to your online account to display your discounted pricing

Dimethylglyoxime, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$333.18 - $957.27

Chemical Identifiers

Chemical Name or Material Dimethylglyoxime
CAS 95-45-4
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3C(:NOH)C(:NOH)CH3
Formula Weight 116.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002747

 
avantor performance
193804
Poly Bottle 125g Each for $333.18

02002748

 
avantor performance
193801
Poly Bottle 500g Each for $957.27
Description & Specifications

Specifications

Formula Weight 116.12
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Dimethylglyoxime
Molecular Formula CH3C(:NOH)C(:NOH)CH3
CAS 95-45-4
Quantity 125g
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 1.37kg