Learn More
Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals
Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19
Supplier: Thermo Scientific Chemicals 408342500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylglyoxime, disodium salt octahydrate | |
75006-64-3 | |
White to Beige | |
Authentic | |
Glass bottle | |
CH3C(NONa)C(NONa)CH3·8H2O | |
250 g | |
(50mg/ml,50% ethanol) Clear solution | |
O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-] | |
304.20 | |
304.19 | |
Crystalline Powder |
99% | |
>300.0°C | |
46% to 49% | |
99% | |
C4H22N2Na2O10 | |
MFCD00150288 | |
dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate | |
IJJDRGUPRATWQX-UHFFFAOYSA-L | |
disodium;2-N,3-N-dioxidobutane-2,3-diimine;octahydrate | |
71310106 | |
99% |
Chemical Identifiers
75006-64-3 | |
304.20 | |
IJJDRGUPRATWQX-UHFFFAOYSA-L | |
71310106 |
C4H22N2Na2O10 | |
MFCD00150288 | |
dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate | |
O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-] |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 262-523-2
TSCA : TSCA
RUO – Research Use Only