Please login to your online account to display your discounted pricing

Dimethylglyoxime, ACS Grade, LabChem™

$88.51 - $88.51

Chemical Identifiers

Chemical Name or Material Dimethylglyoxime
CAS 95-45-4
CAS Max % 100
Purity Grade ACS Grade
Linear Formula CH3C(=NOH)C(=NOH)CH3
Formula Weight 116.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC135969

 
lab chem
LC135969
Poly Bottle 100g Each for $88.51
Description & Specifications

Specifications

Formula Weight 116.12
Purity Grade ACS Grade
CAS Max % 100
Identification Passes Test
Quantity 100g
Physical Form Powder
Linear Formula CH3C(=NOH)C(=NOH)CH3
Melting Point 240°C
Color White
Suitability Passes Test
EINECS Number 202-420-1
Storage Information Room Temperature
Health Hazard 2 GHS H Statement
May form combustible dust concentrations in air.
Chemical Name or Material Dimethylglyoxime
Health Hazard 1 Warning
Packaging Poly Bottle
Solubility Information Soluble in water
CAS 95-45-4
Safety and Handling

Warning

GHS H Statement
May form combustible dust concentrations in air.